Learn More
Growcells IPTG DIOXANE FREE
Supplier:  Growcells MESP2002
Encompass_Preferred
Isopropyltio-beta-D-thiogalctopyronoside is an inducer of beta-galactosidase activity in bacteria. Genomic grade powder.
Catalog No. 50-103-4893