Learn More
Bachem Isopropyl-1-thio-b-D-galactopyranoside (dioxane free)
SDP
Supplier:  Bachem 4026913.0005
Encompass_Preferred
IPTG (dioxane free) , Isopropyl-1-thio-b-D-galactopyranoside (dioxane free), 367-93-1, C9H18O5S, Mr 238.31, 5g. IPTG is an analog of galactose which is not cleaved by E.coli b-galactosidase. Inducer of the lac operon. b-Galactosidase-positive colonies can be detected by X-Gal (5-Bromo-4-chloroindol-3-yl-b-D-galactoside.
Catalog No. 50-260-650