Learn More
Rockland Immunochemicals IPTG (isopropyl beta-D-thiogalactoside) INDUCER for BETA-GALACTOSIDASE EXPRESSION - IPTG-1000
SDP
Catalog No. 501058948
Encompass_Preferred
IPTG (isopropyl beta-D-thiogalactoside) Inducer for Beta-Galactosidase Expression acts as a molecular mimic of a lactose metabolite. The presence of IPTG triggers the activation of the lac operon for downstream gene transcription due to its binding the lac repressor. Due to the presence of a sulfur atom in IPTG, the cells cannot degrade the inducer and therefore its concentration remains constant. IPTG (isopropyl beta-D-thiogalactoside) Inducer for Beta-Galactosidase Expression is ideal for investigators in Immunology, Cell Biology, and Microbiology research.
Catalog No. 50-105-8948 Supplier Rockland Immunochemicals Supplier No. IPTG1000
IPTG (isopropyl beta-D-thiogalactoside) Inducer for Beta-Galactosidase Expression acts as a molecular mimic of a lactose metabolite. The presence of IPTG triggers the activation of the lac operon for downstream gene transcription due to its binding the lac repressor. Due to the presence of a sulfur atom in IPTG, the cells cannot degrade the inducer and therefore its concentration remains constant. IPTG (isopropyl beta-D-thiogalactoside) Inducer for Beta-Galactosidase Expression is ideal for investigators in Immunology, Cell Biology, and Microbiology research.