Learn More
Alkali Scientific IPTG (Isopropyl β D thiogalactopyranoside), Dioxane Free, 200 G
SDP
Catalog No. 5000659784
Encompass_Preferred
IPTG (Isopropyl β-D-thiogalactopyranoside), Dioxane Free, is a widely used chemical reagent in molecular biology, particularly in protein expression systems. This 200g pack is dioxane-free, making it a safer alternative for experiments sensitive to solvent contaminants. IPTG is commonly used to induce gene expression in bacterial systems, particularly in E. coli, by binding to the lac repressor protein and allowing the activation of the lac operon. This product is essential for the study of gene regulation, protein production, and the functional analysis of recombinant proteins. IPTG is ideal for biochemical assays, protein purification, and other research applications.
Catalog No. 50-006-59784 Supplier Alkali Scientific Supplier No. IPT003CP
IPTG (Isopropyl β-D-thiogalactopyranoside), Dioxane Free, is a widely used chemical reagent in molecular biology, particularly in protein expression systems. This 200g pack is dioxane-free, making it a safer alternative for experiments sensitive to solvent contaminants. IPTG is commonly used to induce gene expression in bacterial systems, particularly in E. coli, by binding to the lac repressor protein and allowing the activation of the lac operon. This product is essential for the study of gene regulation, protein production, and the functional analysis of recombinant proteins. IPTG is ideal for biochemical assays, protein purification, and other research applications.