Learn More
Alkali Scientific IPTG (Isopropyl β D thiogalactopyranoside), Dioxane Free, 25 G
SDP
Catalog No. 5000659705
Encompass_Preferred
IPTG (Isopropyl β-D-thiogalactopyranoside), Dioxane Free, 25g, is a molecular biology reagent widely used to induce gene expression in bacterial systems. This dioxane-free formulation is especially beneficial for experiments where the presence of toxic solvents may interfere with the results. IPTG functions by binding to the lac repressor, triggering the expression of lac operon-controlled genes, a common technique in protein expression studies. The 25g packaging is ideal for smaller-scale research or laboratories with limited usage requirements. This compound is essential for recombinant protein production, gene expression analysis, and other molecular biological studies.
Catalog No. 50-006-59705 Supplier Alkali Scientific Supplier No. IPT004CP
IPTG (Isopropyl β-D-thiogalactopyranoside), Dioxane Free, 25g, is a molecular biology reagent widely used to induce gene expression in bacterial systems. This dioxane-free formulation is especially beneficial for experiments where the presence of toxic solvents may interfere with the results. IPTG functions by binding to the lac repressor, triggering the expression of lac operon-controlled genes, a common technique in protein expression studies. The 25g packaging is ideal for smaller-scale research or laboratories with limited usage requirements. This compound is essential for recombinant protein production, gene expression analysis, and other molecular biological studies.