Learn More
Bioworld IPTG Solution, Ready-To-Use Spray, 20 mL
Catalog No. 50198933
Encompass_Preferred
IPTG (Isopropyl-β-D-Thiogalactopyranoside) is an inducer of β-galactosidase. It is used in conjunction with X-gal to promote the expression of proteins in cells controlled by the lac operator system. The solution contains 40mg/ml of IPTG (Isopropyl-β-D-Thiogalactopyranoside) supplied in two very easy and convenient to use format: i. Dropper Bottle (cat# 21530104-1) and ii. Spray Bottle (cat# 21530105-1). Spray bottle: i. Spray (2-3 times) on desired bacterial plateii. Ensure absorption (wait ~5-10 minutes) and iii. Plate the transformed bacteria to be tested and incubateStability: Stable for 6 months at 4°C. It is shipped at ambient temperature.This product is sold for laboratory research use only.
Catalog No. 50-198-933 Supplier Bioworld Supplier No. 215301051
IPTG (Isopropyl-β-D-Thiogalactopyranoside) is an inducer of β-galactosidase. It is used in conjunction with X-gal to promote the expression of proteins in cells controlled by the lac operator system. The solution contains 40mg/ml of IPTG (Isopropyl-β-D-Thiogalactopyranoside) supplied in two very easy and convenient to use format: i. Dropper Bottle (cat# 21530104-1) and ii. Spray Bottle (cat# 21530105-1). Spray bottle: i. Spray (2-3 times) on desired bacterial plateii. Ensure absorption (wait ~5-10 minutes) and iii. Plate the transformed bacteria to be tested and incubateStability: Stable for 6 months at 4°C. It is shipped at ambient temperature.This product is sold for laboratory research use only.