Learn More
Gold Biotechnology Inc IPTG 500 g
SDP
Catalog No. NC1845799
Encompass_Preferred
IPTG is an analog of galactose that is non-metabolizable and inactivates the lac repressor to induce synthesis of β-galactosidase in E. coli. The expression of cloned genes under the control of the lac operon is induced by IPTG. It is also a substrate for thigalactoside transacetylase and has been reported to induce penicillinase in bacteria.
Catalog No. NC1845799 Supplier Gold Biotechnology Inc Supplier No. I2481C500
IPTG is an analog of galactose that is non-metabolizable and inactivates the lac repressor to induce synthesis of β-galactosidase in E. coli. The expression of cloned genes under the control of the lac operon is induced by IPTG. It is also a substrate for thigalactoside transacetylase and has been reported to induce penicillinase in bacteria.