Learn More
Bioworld IPTG (Isopropyl-B-D-ThioGalactopyranoside) Solution, 100mM, 1 mL
Catalog No. 50198868
Encompass_Preferred
Isopropyl-β-D-Thiogalactopyranoside, an inducer of β-galactosidase is used to promote the expression of proteins in cells controlled by the lac operator system. Supplied as 1ml or 10x1ml of 100mM ready-to-use solution to be used with X-gal for blue white colony screening. Techncial Information:Biotechnology Grade.Ready-to-use solution Sterile filtered (0.22μM)Dioxane freeApplication: Petriplates: Use 40 μl per Petriplates (9 cm diameter). LB agar: Use 1μl/mL of liquid LB agar medium just before pouring (~50°C).Liquid culture: Use 1μl/mL of liquid culture medium.Sold as 1mL or 10 x 1mL.
Catalog No. 50-198-868 Supplier Bioworld Supplier No. 215300581
Isopropyl-β-D-Thiogalactopyranoside, an inducer of β-galactosidase is used to promote the expression of proteins in cells controlled by the lac operator system. Supplied as 1ml or 10x1ml of 100mM ready-to-use solution to be used with X-gal for blue white colony screening. Techncial Information:Biotechnology Grade.Ready-to-use solution Sterile filtered (0.22μM)Dioxane freeApplication: Petriplates: Use 40 μl per Petriplates (9 cm diameter). LB agar: Use 1μl/mL of liquid LB agar medium just before pouring (~50°C).Liquid culture: Use 1μl/mL of liquid culture medium.Sold as 1mL or 10 x 1mL.