Learn More
Bioworld IPTG, 2 g
Catalog No. 50198863
Encompass_Preferred
IPTG Isopropyl-β-D-Thiogalactopyranoside. An inducer of β-galactosidase used to promote the expression of proteins in cells controlled by the lac operator system. For blue/white screening, 0.1mM final IPTG concentration in LB (Luria Broth) media is recommended. Techncial Information:Biotechnology Grade.Dioxane free.
Catalog No. 50-198-863 Supplier Bioworld Supplier No. 215300572
IPTG Isopropyl-β-D-Thiogalactopyranoside. An inducer of β-galactosidase used to promote the expression of proteins in cells controlled by the lac operator system. For blue/white screening, 0.1mM final IPTG concentration in LB (Luria Broth) media is recommended. Techncial Information:Biotechnology Grade.Dioxane free.