Learn More
Selleck Chemical LLC IQ-1
SDP
Catalog No. 5000002980
Encompass_Preferred
IQ-1 enables Wnt/ -catenin-driven expansion of mouse ESCs and prevents spontaneous differentiation Maintains embryonic stem cell (ESC) pluripotency in Wnt3a-treated cells and prevents PP2A/Nkd interaction
Catalog No. 50-000-02980 Supplier Selleck Chemical LLC Supplier No. S824810MG
IQ-1 enables Wnt/ -catenin-driven expansion of mouse ESCs and prevents spontaneous differentiation Maintains embryonic stem cell (ESC) pluripotency in Wnt3a-treated cells and prevents PP2A/Nkd interaction