Learn More
Abcam IQ-1, Wnt pathway activator, 1MG
Catalog No. 502706346
Encompass_Preferred
MW 364.4 Da, Purity >95%. Cell-permeable Wnt pathway activator. Targets PR72/130 subunit of PP2A. Maintains ESC pluripotency when co-administered with Wnt3a in vitro. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6346 Supplier Abcam Supplier No. AB1420791MG
MW 364.4 Da, Purity >95%. Cell-permeable Wnt pathway activator. Targets PR72/130 subunit of PP2A. Maintains ESC pluripotency when co-administered with Wnt3a in vitro. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement