Learn More
Selleck Chemical LLC IQ-1S
SDP
Catalog No. 5000002766
Encompass_Preferred
IQ-1S is a JNK3 inhibitor with Kd values of 87 360 and 390 nM for JNK3 JNK2 and JNK1 respectively
Catalog No. 50-000-02766 Supplier Selleck Chemical LLC Supplier No. S67115MG
IQ-1S is a JNK3 inhibitor with Kd values of 87 360 and 390 nM for JNK3 JNK2 and JNK1 respectively