Learn More
Medchemexpress LLC HY-100233 5mg Medchemexpress, IQ-1S (free acid) CAS:23146-22-7 Purity:>98%
SDP
Catalog No. 501931768
Encompass_Preferred
Medchemexpress, HY-100233 5mg IQ-1S (free acid) CAS:23146-22-7 IQ-1S free acid is a prospective inhibitor of NF-κB /activating protein 1 ( AP-1 ) activity with an IC 50 of 2.3±0.41 μM. IQ-1S free acid has binding affinity (K d values) in the nanomolar range for all three JNK s with K d s of 100 nM, 240 nM, and 360 nM for JNK3 , JNK1 , and JNK2 , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1768 Supplier Medchemexpress LLC Supplier No. HY1002335MG
Medchemexpress, HY-100233 5mg IQ-1S (free acid) CAS:23146-22-7 IQ-1S free acid is a prospective inhibitor of NF-κB /activating protein 1 ( AP-1 ) activity with an IC 50 of 2.3±0.41 μM. IQ-1S free acid has binding affinity (K d values) in the nanomolar range for all three JNK s with K d s of 100 nM, 240 nM, and 360 nM for JNK3 , JNK1 , and JNK2 , respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.