Learn More
Bio Rad Laboratories IQ MULTIPLEX POWERMIX
Catalog No. 509012369
Encompass
iQ Multiplex Powermix; Bio-Rad; 50 x 50uL reactions, 2X mix contains dNTPs, 11mM MgCl2, iTaq DNA polymerase, stabilizers
Catalog No. 50-901-2369 Supplier Bio Rad Laboratories Supplier No. 1725848
iQ Multiplex Powermix; Bio-Rad; 50 x 50uL reactions, 2X mix contains dNTPs, 11mM MgCl2, iTaq DNA polymerase, stabilizers