Learn More
Amsco Scientific Steris IQ/OQ 630 STM STETLZR PHSE 2
Catalog No. NC2544409
Encompass
IQ/OQ - Lab Series Steam Sterilizer (Part 2 - Execution)The Execution of a Customer’s pre-approved STERIS IQOQ documentationpackage (ordered separately) is performed on-site by a STERISrepresentative. The Execution follows the IQOQ Document to validate properequipment operation and includes a full NIST or ISO traceable calibration.The STERIS Calibration service will be performed on appropriate processmeasuring equipment used in manufacturing and manufacturing-supportenvironments. Qualified STERIS Service personnel using NIST or ISOtraceable instrumentation perform all program services according todocumented STERIS protocols and procedures. The labor and materialsrequired to execute the procedures/tests of the Part 1 – Document areincluded in this product.THIS SERVICE MUST BE PURCHASED IN CONJUNCTION WITH ACORRESPONDING STERIS IQ/OQ DOCUMENTATION.• IQ/OQ• LIFE SCIENCE1 6,051.00 6,051.00IQ/
Catalog No. NC2544409 Supplier Amsco Scientific Steris Supplier No. SE6001924098
Add to Cart
Add to Cart
Request a Quote
IQ/OQ - Lab Series Steam Sterilizer (Part 2 - Execution)The Execution of a Customer’s pre-approved STERIS IQOQ documentationpackage (ordered separately) is performed on-site by a STERISrepresentative. The Execution follows the IQOQ Document to validate properequipment operation and includes a full NIST or ISO traceable calibration.The STERIS Calibration service will be performed on appropriate processmeasuring equipment used in manufacturing and manufacturing-supportenvironments. Qualified STERIS Service personnel using NIST or ISOtraceable instrumentation perform all program services according todocumented STERIS protocols and procedures. The labor and materialsrequired to execute the procedures/tests of the Part 1 – Document areincluded in this product.THIS SERVICE MUST BE PURCHASED IN CONJUNCTION WITH ACORRESPONDING STERIS IQ/OQ DOCUMENTATION.• IQ/OQ• LIFE SCIENCE1 6,051.00 6,051.00IQ/