Learn More
Amsco Scientific Steris IQ/OQ 630LS STM STETLZR PHSE1
Catalog No. NC2544406
Encompass
IQ/OQ - Lab Series Steam Sterilizer (Part 1 - Document)The STERIS Installation Qualification/Operational Qualification (IQOQ) Serviceis designed to provide the documentation and testing necessary to complywith the appropriate regulatory requirements for validation. This serviceincludes a detailed equipment and site-specific document. This Part 1 -Document requires the Part 2 - Execution to be purchased additionally.Modifications to the structure and/or content of our standard documentationare available for an additional charge. STERIS requires that a minimum ofthree (3) weeks’ notice be given to the responsible Service Manager to allowfor scheduling of personnel. Prior to commencing with this service, STERISrequires that the equipment be fully started-up and operational. In addition,all of the necessary utilities must be available and operational withoutinterruption for the duration of the IQOQ.
Catalog No. NC2544406 Supplier Amsco Scientific Steris Supplier No. SE6001924
Add to Cart
Add to Cart
Request a Quote
IQ/OQ - Lab Series Steam Sterilizer (Part 1 - Document)The STERIS Installation Qualification/Operational Qualification (IQOQ) Serviceis designed to provide the documentation and testing necessary to complywith the appropriate regulatory requirements for validation. This serviceincludes a detailed equipment and site-specific document. This Part 1 -Document requires the Part 2 - Execution to be purchased additionally.Modifications to the structure and/or content of our standard documentationare available for an additional charge. STERIS requires that a minimum ofthree (3) weeks’ notice be given to the responsible Service Manager to allowfor scheduling of personnel. Prior to commencing with this service, STERISrequires that the equipment be fully started-up and operational. In addition,all of the necessary utilities must be available and operational withoutinterruption for the duration of the IQOQ.