Learn More
IQ/OQ/PQ Kit, For Ion Chromatography, Certified Reference Material, MilliporeSigma™ Supelco™
Catalog No. 111003894 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
11-100-3894 1 Kit
1 options
Catalog No. 11-100-3894 Supplier MilliporeSigma Supelco Supplier No. 126741KT
Add to Cart
Add to Cart
IQ/OQ/PQ-Kit for the qualification of ion chromatography systems. Suitable for the verification of linearity, repeatability and carry over. Contains six certified bromide standards (5, 10, 20, 50, 100 und 1000 mg/kg; 100 mL each). The exact contents, traceability and uncertainty are given in a detailed certificate. This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034.. This CRM is traceable to SI unit kg and measured against primary material from an NMI, e.g. NIST.Part of the "IQ/OQ Kit for IC" from Metrohm AG, part number 6.5333.000
TRUSTED_SUSTAINABILITY
Specifications
Quantity 1 Kit
Grade Certified Reference Material