Learn More
Aviva Systems Biology Corp IQCD Antibody - N-terminal region (ARP52610_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52610_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IQCD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human IQCD. Alternate names: 4933433C09RikApplicationWB. Species reactivity: Human
Catalog No. 50-014-60784