Learn More
Aviva Systems Biology Corp IQCD Antibody - middle region (ARP52611_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52611_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against IQCD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human IQCD. Alternate names: 4933433C09RikApplicationWB. Species reactivity: Dog, Human, Rat, Yeast
Catalog No. 50-285-740