Learn More
Abcam IQDMA, Cell-permeable STAT5 inhibitor, 10MG
Catalog No. 502706816
Encompass_Preferred
MW 304.39 g/mol, Purity >98%. Cell-permeable STAT5 inhibitor (IC₅₀ = 8 μM). Indoloquinoline derivative. Antineoplastic agent with broad spectrum antitumor activity seen in vitro. Induces cell-cycle arrest at the G₂/M phase.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6816 Supplier Abcam Supplier No. AB14119210MG
MW 304.39 g/mol, Purity >98%. Cell-permeable STAT5 inhibitor (IC₅₀ = 8 μM). Indoloquinoline derivative. Antineoplastic agent with broad spectrum antitumor activity seen in vitro. Induces cell-cycle arrest at the G₂/M phase.
The product is subject to the following: Abcam Restricted Use Statement