Learn More
Broadpharm IR 750 NHS Ester, 100 mg, near-infrared fluorescent dye, water-soluble, NHS ester reactive group, pH-insensitive, stable signals, suitable for flow cytometry and microscopy
Catalog No. NC3490796
Encompass
IR 750 NHS Ester, 100 mg, near-infrared fluorescent dye, water-soluble, NHS ester reactive group, pH-insensitive, stable signals, suitable for flow cytometry and microscopy
Catalog No. NC3490796 Supplier Broadpharm Supplier No. BP28167100MG
IR 750 NHS Ester, 100 mg, near-infrared fluorescent dye, water-soluble, NHS ester reactive group, pH-insensitive, stable signals, suitable for flow cytometry and microscopy