Learn More
TARGETMOL CHEMICALS INC IR-780 IODIDE 25MG
SDP
Catalog No. 502577228
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IR-780 Iodide (IR 780 iodide) a near‐infrared fluorescent dye is used for the exclusive characterization of human CSCs through the HIF‐1(alpha)/glycolysis dependent mitochondrial transporter ABCB10's activity. purity: 99%
Catalog No. 50-257-7228 Supplier TARGETMOL CHEMICALS INC Supplier No. T2097425MG
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. IR-780 Iodide (IR 780 iodide) a near‐infrared fluorescent dye is used for the exclusive characterization of human CSCs through the HIF‐1(alpha)/glycolysis dependent mitochondrial transporter ABCB10's activity. purity: 99%