Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC IR-783 | 115970-66-6 | 99.9% | 749.35 | 250 MG
SDP

Catalog No. 5000352290
Encompass_Preferred

IR-783 is a near-infrared (NIR) heptamethine cyanine fluorescent probe. It has been shown to significantly inhibit tumor growth and induce apoptosis in MDA-MB-231 xenograft models, making it suitable for breast cancer research.

  • Near-infrared heptamethine cyanine fluorescent probe
  • Inhibits tumor growth
  • Induces apoptosis in MDA-MB-231 xenograft models
  • Suitable for breast cancer research
  • Purity: 99.9%
  • Molecular weight: 749.35
  • Appearance: Green to dark green solid

Catalog No. 50-003-52290 Supplier Medchemexpress LLC Supplier No. 250MGHY139109
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

IR-783 is a near-infrared (NIR) heptamethine cyanine fluorescent probe. It has been shown to significantly inhibit tumor growth and induce apoptosis in MDA-MB-231 xenograft models, making it suitable for breast cancer research.

  • Near-infrared heptamethine cyanine fluorescent probe
  • Inhibits tumor growth
  • Induces apoptosis in MDA-MB-231 xenograft models
  • Suitable for breast cancer research
  • Purity: 99.9%
  • Molecular weight: 749.35
  • Appearance: Green to dark green solid

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.