Learn More
Medchemexpress LLC IR-783 (ADS 780WS) | 115970-66-6 | 99.9% | 749.35 | 1 ML
SDP
Catalog No. 5000348356
Encompass_Preferred
IR-783 (ADS 780WS) is a near-infrared (NIR) heptamethine cyanine fluorescent probe that significantly inhibits tumor growth and induces apoptosis in the MDA-MB-231 xenograft model. It is suitable for breast cancer research.
  • Functions as a near-infrared heptamethine cyanine fluorescent probe.
  • Demonstrates significant inhibition of tumor growth.
  • Induces apoptosis in a xenograft model.
  • Useful for breast cancer studies.
Catalog No. 50-003-48356 Supplier Medchemexpress LLC Supplier No. HY13910910MM/1ML
IR-783 (ADS 780WS) is a near-infrared (NIR) heptamethine cyanine fluorescent probe that significantly inhibits tumor growth and induces apoptosis in the MDA-MB-231 xenograft model. It is suitable for breast cancer research.
  • Functions as a near-infrared heptamethine cyanine fluorescent probe.
  • Demonstrates significant inhibition of tumor growth.
  • Induces apoptosis in a xenograft model.
  • Useful for breast cancer studies.