Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich IR-792 perchlorate

Catalog No. 4259821G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-598-21G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 42-598-21G Supplier Sigma Aldrich Supplier No. 4259821G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

IR-792 perchlorate

Specifications

CAS 207399-10-8
Melting Point 201°C to 203°C
Molecular Formula C42H49ClN2O4S
Linear Formula C42H49ClN2O4S
MDL Number MFCD00467006
Quantity 1 g
Synonym 2-[2-[3-[(1,3-Dihydro-3,3-dimethyl-1-propyl-2H-indol-2-ylidene)ethylidene]-2-(phenylthio)-1-cyclohexen-1-yl]ethenyl]-3,3-dimethyl-1-propylindolium perchlorate
Molecular Weight (g/mol) 713.37
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.