Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IR-813 p-Toluenesulfonate 98.0+%, TCI America™
SDP

Catalog No. C28865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C28865G 5 g
C28861G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C28865G Supplier TCI America Supplier No. C28865G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 134127-48-3
Molecular Formula C47H47ClN2O3S
Molecular Weight (g/mol) 755.41
MDL Number MFCD03093252
InChI Key LQVQHHTYXSPHSQ-UHFFFAOYSA-M
Synonym 2-[2-[2-Chloro-3-[2-(1,3-dihydro-1,1,3-trimethyl-2H-benz[e]indol-2-ylidene)ethylidene]-1-cyclohexen-1-yl]ethenyl]-1,1,3-trimethyl-1H-benz[e]indolium 4-Methylbenzenesulfonate, 2-Chloro-3-[2-(1,1,3-trimethylbenz[e]indolin-2-ylidene)ethylidene]-1-[2-(1,1,3-t
PubChem CID 91972145
IUPAC Name 2-{2-[2-chloro-3-(2-{1,1,3-trimethyl-1H,2H,3H-benzo[e]indol-2-ylidene}ethylidene)cyclohex-1-en-1-yl]ethenyl}-1,1,3-trimethyl-1H-benzo[e]indol-3-ium 4-methylbenzene-1-sulfonate
SMILES CC1=CC=C(C=C1)S([O-])(=O)=O.CN1C2=CC=C3C=CC=CC3=C2C(C)(C)C1=CC=C1CCCC(C=CC2=[N+](C)C3=CC=C4C=CC=CC4=C3C2(C)C)=C1Cl

Specifications

Quantity 5 g
Formula Weight 755.41
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material IR-813 p-Toluenesulfonate
EINECSNumber (5)-6789
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.