Learn More
Medchemexpress LLC IR 813 tosylate | 134127-48-3 | MFCD03093252 | 99.5% | 755.40 g/mol | C47H47ClN2O3S | 250 MG
SDP
Catalog No. 5000281350
Encompass_Preferred
IR 813 tosylate is a near-infrared (NIR) fluorescent dye for in vivo imaging applications, commonly used to visualize regional lymph nodes in mice. It is supplied as a high-purity solid with recommended storage and solvent handling instructions for research use.
  • High purity (99.47%) suitable for research imaging.
  • Near-infrared emission for improved tissue penetration and contrast.
  • Soluble in DMSO (62.5 mg/mL) with ultrasonic assistance.
  • Store solid at 4°C, sealed, protected from moisture and light.
  • Solution stability: -80°C up to 6 months, -20°C up to 1 month.
  • Supplied in small research quantities for laboratory studies.
Catalog No. 50-002-81350 Supplier Medchemexpress LLC Supplier No. HYW248594250MG
IR 813 tosylate is a near-infrared (NIR) fluorescent dye for in vivo imaging applications, commonly used to visualize regional lymph nodes in mice. It is supplied as a high-purity solid with recommended storage and solvent handling instructions for research use.
  • High purity (99.47%) suitable for research imaging.
  • Near-infrared emission for improved tissue penetration and contrast.
  • Soluble in DMSO (62.5 mg/mL) with ultrasonic assistance.
  • Store solid at 4°C, sealed, protected from moisture and light.
  • Solution stability: -80°C up to 6 months, -20°C up to 1 month.
  • Supplied in small research quantities for laboratory studies.