Learn More
Medchemexpress LLC Ir 813 tosylate | 134127-48-3 | MFCD03093252 | 99.5% | 755.41 g/mol | C47H47ClN2O3S | 500 MG
SDP
Catalog No. 5000282130
Encompass_Preferred
IR 813 tosylate is a near-infrared (NIR) fluorescent dye for research use in biological imaging. It generates a strong NIR signal suitable for in vivo experiments and is commonly used to visualize regional lymph nodes in small-animal models. The material is supplied with supporting analytical and safety documentation to aid reproducible and safe use.
  • Near-infrared fluorescence suitable for in vivo imaging.
  • High purity (reported ~99.5%) for reproducible results.
  • Supplied as a stable solid for straightforward storage and handling.
  • Provided with COA, SDS, and spectral/analytical data.
  • Compatible with common NIR imaging systems and filters.
  • Useful for lymph node visualization in small-animal studies.
Catalog No. 50-002-82130 Supplier Medchemexpress LLC Supplier No. HYW248594500MG
IR 813 tosylate is a near-infrared (NIR) fluorescent dye for research use in biological imaging. It generates a strong NIR signal suitable for in vivo experiments and is commonly used to visualize regional lymph nodes in small-animal models. The material is supplied with supporting analytical and safety documentation to aid reproducible and safe use.
  • Near-infrared fluorescence suitable for in vivo imaging.
  • High purity (reported ~99.5%) for reproducible results.
  • Supplied as a stable solid for straightforward storage and handling.
  • Provided with COA, SDS, and spectral/analytical data.
  • Compatible with common NIR imaging systems and filters.
  • Useful for lymph node visualization in small-animal studies.