Learn More
Medchemexpress LLC Ir-820 (new indocyanine green) | 172616-80-7 | 96.36% | 849.47 | 250 MG
SDP
Catalog No. 5000347833
Encompass_Preferred
IR-820 (New Indocyanine Green) is an infrared blood pool contrast agent. It is also commonly used as a laser and near-infrared dye for detecting and quantifying diseased tissue in live animals.
  • Maximal excitation wavelength of 710 nm
  • Maximal emission wavelength of 820 nm
  • Appearance as a solid, brown to black substance
  • Functions as an infrared blood pool contrast agent
  • Serves as a laser and near-infrared dye for detecting and quantifying diseased tissue in living organisms
Catalog No. 50-003-47833 Supplier Medchemexpress LLC Supplier No. 250MGHY136886
Add to Cart
Add to Cart
IR-820 (New Indocyanine Green) is an infrared blood pool contrast agent. It is also commonly used as a laser and near-infrared dye for detecting and quantifying diseased tissue in live animals.
  • Maximal excitation wavelength of 710 nm
  • Maximal emission wavelength of 820 nm
  • Appearance as a solid, brown to black substance
  • Functions as an infrared blood pool contrast agent
  • Serves as a laser and near-infrared dye for detecting and quantifying diseased tissue in living organisms