Learn More
Medchemexpress LLC IR-825 | 1558079-49-4 | 98.3% | 824.42 | C54H48ClN2O4 | 25 MG
SDP
Catalog No. 5000229879
Encompass_Preferred
IR-825 is a near-infrared fluorescent dye with a terminal carboxyl (COOH) functional group that enables covalent conjugation to other molecules. It is used in research applications such as near-infrared imaging and tumor photothermal treatment. The compound has formula C54H48ClN2O4 and a molecular weight of 824.42 Da.
  • Near-infrared fluorescent dye suitable for imaging and photothermal applications
  • Terminal carboxyl (COOH) group for covalent conjugation to biomolecules
  • High purity for research use (listed as 98.27%)
  • Molecular weight 824.42 Da and formula C54H48ClN2O4
Catalog No. 50-002-29879 Supplier Medchemexpress LLC Supplier No. HY11892425MG
IR-825 is a near-infrared fluorescent dye with a terminal carboxyl (COOH) functional group that enables covalent conjugation to other molecules. It is used in research applications such as near-infrared imaging and tumor photothermal treatment. The compound has formula C54H48ClN2O4 and a molecular weight of 824.42 Da.
  • Near-infrared fluorescent dye suitable for imaging and photothermal applications
  • Terminal carboxyl (COOH) group for covalent conjugation to biomolecules
  • High purity for research use (listed as 98.27%)
  • Molecular weight 824.42 Da and formula C54H48ClN2O4