Learn More
Medchemexpress LLC Ir-825 | 1558079-49-4 | 98.3% | 824.42 | C54H48ClN2O4 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY11892450MG
Encompass_Preferred
IR-825 is a near-infrared fluorescent dye with a terminal carboxyl (COOH) functional group that allows covalent conjugation to other molecules. It is supplied as a purified solid for research use in imaging and photothermal therapy applications.
  • Near-infrared fluorescent dye suitable for imaging and photothermal studies.
  • Carboxyl (COOH) functional group enables conjugation to biomolecules.
  • High purity suitable for research: 98.27%.
  • Molecular weight 824.42; formula C54H48ClN2O4.
  • Available in small lab quantities, including 50 mg packs.
Catalog No. 50-002-27500