Learn More
Medchemexpress LLC IR-825 | 1558079-49-4 | MFCD32068082 | 98.3% | 824.42 g/mol | C54H48ClN2O4 | 10 MG
SDP
Catalog No. 5000308089
Encompass_Preferred
IR-825 is a near-infrared fluorescent dye with a terminal carboxyl (COOH) functional group that enables covalent conjugation to other molecules. It is supplied as a high-purity research reagent suitable for near-infrared imaging, fluorescent labeling, and tumor photothermal treatment studies.
  • Near-infrared fluorescent dye suitable for imaging and photothermal research.
  • Terminal carboxyl (COOH) group enables covalent conjugation to biomolecules and probes.
  • High purity (98.3%) for research applications.
  • Molecular weight ≈ 824.42 g/mol; formula C54H48ClN2O4.
  • Available in small, weighed packages for laboratory use (for example, 10 MG).
Catalog No. 50-003-08089 Supplier Medchemexpress LLC Supplier No. HY11892410MG
IR-825 is a near-infrared fluorescent dye with a terminal carboxyl (COOH) functional group that enables covalent conjugation to other molecules. It is supplied as a high-purity research reagent suitable for near-infrared imaging, fluorescent labeling, and tumor photothermal treatment studies.
  • Near-infrared fluorescent dye suitable for imaging and photothermal research.
  • Terminal carboxyl (COOH) group enables covalent conjugation to biomolecules and probes.
  • High purity (98.3%) for research applications.
  • Molecular weight ≈ 824.42 g/mol; formula C54H48ClN2O4.
  • Available in small, weighed packages for laboratory use (for example, 10 MG).