Learn More
International Crystal IR CARDS POT BROMIDE 25PK
Catalog No. NC9994860
Encompass_Preferred
Real Crystal; IR Sample Card, KBr, 19mm Aperture, 2 in. x 4 in., Fit in the Universal Slide Mount of any Spectrophotometer, Do not Exhibit the Strong Absorbance Peaks, Transmit in Excess of 80% of the Available Energy
Catalog No. NC9994860 Supplier International Crystal Supplier No. 00007090
Real Crystal; IR Sample Card, KBr, 19mm Aperture, 2 in. x 4 in., Fit in the Universal Slide Mount of any Spectrophotometer, Do not Exhibit the Strong Absorbance Peaks, Transmit in Excess of 80% of the Available Energy