Learn More
Strem, An Ascensus Company CAS# 870987-63-6. 1g. (4,4'-Di-t-butyl-2,2'-bipyridine)bis[3,5-difluoro-2-[5-trifluoromethyl-2-pyridinyl-kN)phenyl-kC]iridium(III) hexafluorophosphate, 99%.
SDP
Supplier:  Strem, An Ascensus Company 7704251G
Encompass
CAS# 870987-63-6. 1g. (4,4'-Di-t-butyl-2,2'-bipyridine)bis[3,5-difluoro-2-[5-trifluoromethyl-2-pyridinyl-kN)phenyl-kC]iridium(III) hexafluorophosphate, 99%. . Molecular Weight: 1121.91. Molecular Formula: [Ir(C18H24N2)(C12H5F5N)2]+PF6ae. Color/form: yellow xtl. Strem# 77-0425. http://www.strem.com/catalog/v/77-0425/
Catalog No. 50-201-6660