Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich [Ir(Dfcf3Ppy)2-(5,5'-Dcf3Bpy)]Pf6

Catalog No. 908568500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
90-856-8500MG 500 mg
90-856-8100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 90-856-8500MG Supplier Sigma Aldrich Supplier No. 908568500MG
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

[Ir(Dfcf3Ppy)2-(5,5'-Dcf3Bpy)]Pf6

Specifications

CAS 808142-88-3
Molecular Formula C42H42F8IrN4P
Linear Formula C42H42F8IrN4P
Quantity 500 mg
Synonym [(4,4'-di-tert-butyl-2,2'-bipyridine)-bis-(5-methyl-2-(5-fluoro-phenyl)-pyridine)-iridium(III)] hexafluorophosphate
Molecular Weight (g/mol) 977.99 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.