Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich [Ir(Dtbbpy)(Ppy)2]Pf6

Catalog No. 74776950MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
74-776-950MG 50 mg
74-776-9100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 74-776-950MG Supplier Sigma Aldrich Supplier No. 74776950MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

[Ir(dtbbpy)(ppy)2]PF6 is an iridium-based photocatalyst which is used to drive various photochemical reactions such as:Coupling of N-arylamines and nitroalkanes via an oxidative aza-Henry reaction.Atom transfer radical addition (ATRA) of haloalkanes to alkenes and alkynes.Conversion of 2-bromoanilides to the corresponding 3,3-disubstituted oxindoles.Aryl-alkyl C-C reductive cross-coupling reactions.

Specifications

CAS 676525-77-2
Molecular Formula C40H40F6IrN4P
Linear Formula C40H40F6IrN4P
Quantity 50 mg
Synonym [Ir(dtbbpy)(ppy)2][PF6]; Ir Photocatalyst; [4,4'-Bis(1,1-dimethylethyl)-2,2'-bipyridine-N1,N1']bis[2-(2-pyridinyl-N)phenyl-C]iridium(III) hexafluorophosphate
Molecular Weight (g/mol) 913.95 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.