Learn More
Strem, An Ascensus Company [Ir(dtbbpy)(ppy)2]PF6,99%100mg
SDP
Supplier:  Strem, An Ascensus Company 770410100MG
Encompass
CAS# 676525-77-2. 100mg. (4,4'-Di-t-butyl-2,2'-bipyridine)bis[2-(2-pyridinyl-kN)phenyl-kC]iridium(III) hexafluorophosphate, 99%. MFCD28369259. Molecular Weight: 913.95. Molecular Formula: [Ir(C18H24N2)(C11H8N)2]+PF6ae. Color/form: yellow xtl. Strem# 77-0410. http://www.strem.com/catalog/v/77-0410/
Catalog No. 50-203-4202