Learn More
Strem, An Ascensus Company CAS 500295-52-3. 250mg. Tris[(2-(2-pyridinyl-kN)-5-(trifluoromethyl)phenyl-kC]iridium(III) 95%.
SDP
Supplier:  Strem, An Ascensus Company 776580250MG
Encompass
CAS 500295-52-3. 250mg. Tris[(2-(2-pyridinyl-kN)-5-(trifluoromethyl)phenyl-kC]iridium(III) 95%. . Molecular Weight 858.78. Molecular Formula C36H21F9IrN3. Color/form yellow solid Strem 77-6580. http//www.strem.com/catalog/v/77-6580/
Catalog No. 50-211-0860