Learn More
Strem, An Ascensus Company CAS . 100mg. (R)-()-7-[N-(13-dithian-2-yl)methylamino]-7-[bis(35-di-t-butylphenyl)phosphino]-2233-tetrahydro-11-spirobindanechlorodihydroiridium(III) 97% [Ir-(R)-DTB-SpiroSAP].
SDP
Supplier:  Strem, An Ascensus Company 772510100MG
Encompass
100mg. (R)-()-7-[N-(13-dithian-2-yl)methylamino]-7-[bis(35-di-t-butylphenyl)phosphino]-2233-tetrahydro-11-spirobindanechlorodihydroiridium(III) 97% [Ir-(R)-DTB-SpiroSAP]. . Molecular Weight 1005.86. Molecular Formula C50H68ClIrNPS2. Color/form pale yellow solid Strem 77-2510. http//www.strem.com/catalog/v/77-2510/
Catalog No. 50-211-0850