Learn More
Andwin Scientific IR THERMOMETER GUN W/LASER
SDP
Catalog No. NC0616710
Encompass_Preferred
The Traceable Infrared Thermometer Gun features a small, lightweight design that enables users to take temperature readings of any surface in a quick moment. This product features a laser sighting beam that allows direct aim at target surface even f
Catalog No. NC0616710 Supplier Andwin Scientific Supplier No. 12777846
The Traceable Infrared Thermometer Gun features a small, lightweight design that enables users to take temperature readings of any surface in a quick moment. This product features a laser sighting beam that allows direct aim at target surface even f