Learn More
METRIS INSTRUMENTS EAST LLC IR THERMOMETER IRT424-2L 30:1
Catalog No. 50320901
Encompass_Preferred
Thermometer, IR; Metris Instruments; Range: - 60 deg. to 860 deg. C; +/- 2% accuracy; Adjustable Emissivity; D: S: 30: 1; Dual Laser pointers; MIN-MAX-DIF-AVG display; 9x6x2.8 in.; 2 x AAA batt
Catalog No. 50-320-901 Supplier METRIS INSTRUMENTS EAST LLC Supplier No. IRT4242L
Thermometer, IR; Metris Instruments; Range: - 60 deg. to 860 deg. C; +/- 2% accuracy; Adjustable Emissivity; D: S: 30: 1; Dual Laser pointers; MIN-MAX-DIF-AVG display; 9x6x2.8 in.; 2 x AAA batt