Learn More
Grainger IR Thermometer SingleDot -49 to 752F
Catalog No. 502750934
Encompass_Preferred
The Taylor 9842 Commercial Infrared Thermometer is a handheld non-contact infrared thermometer that measures surface temperatures. It has a temperature range of -58°F to 1022°F (-50°C to 550°C) with accuracy of ±2% of reading or ±4°F (±2°C). The unit features a 12:1 distance to spot ratio with dual lasers for targeting. It has a backlit LCD display, auto shut-off, and low battery indicator. The thermometer is powered by a 9V battery. It comes with a carrying case. The Taylor 9842 is designed for HVAC, food service, industrial, commercial, residential and automotive applications.
Catalog No. 50-275-0934 Supplier Grainger Supplier No. 14F320
The Taylor 9842 Commercial Infrared Thermometer is a handheld non-contact infrared thermometer that measures surface temperatures. It has a temperature range of -58°F to 1022°F (-50°C to 550°C) with accuracy of ±2% of reading or ±4°F (±2°C). The unit features a 12:1 distance to spot ratio with dual lasers for targeting. It has a backlit LCD display, auto shut-off, and low battery indicator. The thermometer is powered by a 9V battery. It comes with a carrying case. The Taylor 9842 is designed for HVAC, food service, industrial, commercial, residential and automotive applications.