Learn More
METRIS INSTRUMENTS EAST LLC IR THERMOMETER TN418L1 12:1
Catalog No. 50320897
Encompass_Preferred
Thermometer, IR; Metris Instruments; Temp. range: -60deg. to 500deg.C; +/-2% accuracy; Resolution: 0.1deg.C/F; Fixed emissivity: 0.95; D:S: 12:1; Laser pointer; Max. temp. display; 1.81 x 5.63 x 7.28 in.; 2 x AAA batteries
Catalog No. 50-320-897 Supplier METRIS INSTRUMENTS EAST LLC Supplier No. TN418L1
Thermometer, IR; Metris Instruments; Temp. range: -60deg. to 500deg.C; +/-2% accuracy; Resolution: 0.1deg.C/F; Fixed emissivity: 0.95; D:S: 12:1; Laser pointer; Max. temp. display; 1.81 x 5.63 x 7.28 in.; 2 x AAA batteries