Learn More
METRIS INSTRUMENTS EAST LLC IR THERMOMETER TN418L1 NIST
Catalog No. 50320898
Encompass_Preferred
Thermometer, IR; Metris Instruments; NIST; Temp.: - 60 deg. - 500 deg. C; +/- 2% accuracy; Resolution: 0.1 deg. C/F; Fixed emissivity: 0.95; D: S: 12: 1; Laser p
Catalog No. 50-320-898 Supplier METRIS INSTRUMENTS EAST LLC Supplier No. TN418L1NIST
Thermometer, IR; Metris Instruments; NIST; Temp.: - 60 deg. - 500 deg. C; +/- 2% accuracy; Resolution: 0.1 deg. C/F; Fixed emissivity: 0.95; D: S: 12: 1; Laser p