Learn More
METRIS INSTRUMENTS EAST LLC IR THERMOMETER TN418LD 16:1
Catalog No. 50320899
Encompass_Preferred
Thermometer, IR; Metris Instruments; Range: - 60 deg. to 625 deg. C; +/- 2% accuracy; Adjustable Emissivity; D: S: 16: 1; Laser pointer; MIN-MAX-DIF-AVG display; K-Type thermocouple port; 1.81x
Catalog No. 50-320-899 Supplier METRIS INSTRUMENTS EAST LLC Supplier No. TN418LD
Thermometer, IR; Metris Instruments; Range: - 60 deg. to 625 deg. C; +/- 2% accuracy; Adjustable Emissivity; D: S: 16: 1; Laser pointer; MIN-MAX-DIF-AVG display; K-Type thermocouple port; 1.81x