Learn More
METRIS INSTRUMENTS EAST LLC IR THERMOMETER TN418LD NIST
Catalog No. 50320900
Encompass_Preferred
Thermometer, IR; Metris Instruments; NIST; Range: - 60 deg. to 625 deg. C; +/- 2% accuracy; Adjustable Emissivity; D: S: 16: 1; Laser pointer; MIN-MAX-DIF-AVG display; K-Type thermocouple port;
Catalog No. 50-320-900 Supplier METRIS INSTRUMENTS EAST LLC Supplier No. TN418LDNIST
Thermometer, IR; Metris Instruments; NIST; Range: - 60 deg. to 625 deg. C; +/- 2% accuracy; Adjustable Emissivity; D: S: 16: 1; Laser pointer; MIN-MAX-DIF-AVG display; K-Type thermocouple port;