Learn More
METRIS INSTRUMENTS EAST LLC IR THERMOMETER TN425LE NIST
Catalog No. 50320904
Encompass_Preferred
Thermometer, IR; Metris Instruments; NIST; Range: - 60 deg. to 1000 deg. C; +/- 2% accuracy; Adjustable Emissivity; D: S: 50: 1; Dual Laser pointers; MIN-MAX-DIF-AVG displays; 8x8x2.8 in.; 2 x
Catalog No. 50-320-904 Supplier METRIS INSTRUMENTS EAST LLC Supplier No. TN425LENIST
Thermometer, IR; Metris Instruments; NIST; Range: - 60 deg. to 1000 deg. C; +/- 2% accuracy; Adjustable Emissivity; D: S: 50: 1; Dual Laser pointers; MIN-MAX-DIF-AVG displays; 8x8x2.8 in.; 2 x