Learn More
Cole-Parmer Extech 42510A Compact Infrared Thermometer (12: 1)
Catalog No. 50993116 Shop All Cole-Parmer Products
Encompass
Measures non-contact surface temperature up to 1000F. Compact thermometer with wide temperature range from -58 to 1000F (-50 to 538C). Audible and visible overrange indicators. Built-in laser pointer identifies target area. 8:1 distance to target ratio. Backlighting illuminates display for taking measurements at night or in areas with low background light levels.
Catalog No. 50-993-116 Supplier Cole-Parmer Supplier No. 9041514
Measures non-contact surface temperature up to 1000F. Compact thermometer with wide temperature range from -58 to 1000F (-50 to 538C). Audible and visible overrange indicators. Built-in laser pointer identifies target area. 8:1 distance to target ratio. Backlighting illuminates display for taking measurements at night or in areas with low background light levels.