Learn More
Medchemexpress LLC IR415 100mg | 452967-14-5 | 100 MG
SDP
Catalog No. 5000307548
Encompass_Preferred
IR415 is a small-molecule research inhibitor that blocks hepatitis B virus (HBV) replication by binding HBx and reversing HBx-mediated suppression of RNA interference. It is intended for in vitro studies of HBV replication and HBx biology.
  • Potent HBx binder with a reported Kd of about 2 nM.
  • Useful for in vitro HBV replication and mechanism-of-action studies.
  • High reported chemical purity (99.66%).
  • Molecular weight ≈ 296.34 and formula C13H14F2N4S.
  • Recommended powder storage: -20°C for long-term stability.
Catalog No. 50-003-07548 Supplier Medchemexpress LLC Supplier No. HY116999100MG
IR415 is a small-molecule research inhibitor that blocks hepatitis B virus (HBV) replication by binding HBx and reversing HBx-mediated suppression of RNA interference. It is intended for in vitro studies of HBV replication and HBx biology.
  • Potent HBx binder with a reported Kd of about 2 nM.
  • Useful for in vitro HBV replication and mechanism-of-action studies.
  • High reported chemical purity (99.66%).
  • Molecular weight ≈ 296.34 and formula C13H14F2N4S.
  • Recommended powder storage: -20°C for long-term stability.