Learn More
Medchemexpress LLC IR415 10mg | 452967-14-5 | 296.34 g/mol | C13H14F2N4S | 10 MG
SDP
Catalog No. 5000229805
Encompass_Preferred
IR415 is a small-molecule inhibitor of hepatitis B virus (HBV) that blocks HBx activity and inhibits HBV replication. It selectively interacts with HBx (Kd = 2 nM) and reverses HBx-mediated suppression of dicer endoribonuclease activity. Supplied as a white to off-white solid for research use.
  • Inhibits HBV replication by blocking HBx activity.
  • Selectively binds HBx (Kd = 2 nM).
  • High purity suitable for research applications.
  • Molecular weight 296.34 g/mol and formula C13H14F2N4S.
  • Solid, white to off-white physical form for powder handling.
  • Stable under recommended storage for powdered material.
  • Available in small research quantities and solution formats.
  • Intended for research use only.
Catalog No. 50-002-29805 Supplier Medchemexpress LLC Supplier No. HY11699910MG
IR415 is a small-molecule inhibitor of hepatitis B virus (HBV) that blocks HBx activity and inhibits HBV replication. It selectively interacts with HBx (Kd = 2 nM) and reverses HBx-mediated suppression of dicer endoribonuclease activity. Supplied as a white to off-white solid for research use.
  • Inhibits HBV replication by blocking HBx activity.
  • Selectively binds HBx (Kd = 2 nM).
  • High purity suitable for research applications.
  • Molecular weight 296.34 g/mol and formula C13H14F2N4S.
  • Solid, white to off-white physical form for powder handling.
  • Stable under recommended storage for powdered material.
  • Available in small research quantities and solution formats.
  • Intended for research use only.